4-Hydroxy-3-methoxyphenylacetonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-Hydroxy-3-methoxyphenyl)acetonitrile
OR52500-1G
APOSOR52500-1GEA
139
EUR
APOSOR52500-1G
4-Hydroxy-3-methoxyphenylacetonitrile
4-Hydroxy-3-methoxyphenylacetonitrile
Formel:
C₉H₉NO₂ Molecular Weight: 163.18 g/mol Schmelzpunkt: 56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001920 CAS-Nummer: 4468-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...