4-Chloro-2-iodopyrimidine

Lieferant: Apollo Scientific
Achtung

OR52502-1G
APOSOR52502-1GEA 632 EUR
APOSOR52502-1G
4-Chloro-2-iodopyrimidine
4-Chloro-2-iodopyrimidine
Formel: C₄H₂ClIN₂
Molecular Weight: 240,43 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18411135
CAS-Nummer: 1108165-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR