Methyl 1-methyl-4-nitro-1H-pyrrole-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR52507-1G
APOSOR52507-1GEA 154 EUR
APOSOR52507-1G
Methyl 1-methyl-4-nitro-1H-pyrrole-2-carboxylate
Methyl 1-methyl-4-nitro-1H-pyrrole-2-carboxylate
Formel: C₇H₈N₂O₄
Molecular Weight: 184,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03789112
CAS-Nummer: 13138-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR