Ethyl-1-methyl-4-nitroimidazole-2-carboxylate 98%

Lieferant: Apollo Scientific

OR52509-5G
APOSOR52509-5GEA 84.1 EUR
APOSOR52509-5G
Ethyl-1-methyl-4-nitroimidazole-2-carboxylate 98%
Ethyl-1-methyl-4-nitroimidazole-2-carboxylate
Formel: C₇H₉N₃O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03789110
CAS-Nummer: 109012-23-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR