Ethyl-4-amino-1-methyl-1H-imidazole-2-carboxylate hydrochloride

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-amino-1-methyl-1H-imidazole-2-carboxylate hydrochloride, 4-Amino-1-methyl-1H-imidazole-2-carboxylic acid-ethyl ester·HCl

OR52510-1G
APOSOR52510-1GEA 111 EUR
APOSOR52510-1G
Ethyl-4-amino-1-methyl-1H-imidazole-2-carboxylate hydrochloride
Ethyl-4-amino-1-methyl-1H-imidazole-2-carboxylate hydrochloride
Formel: C₇H₁₂ClN₃O₂
Molecular Weight: 205.642 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01318760
CAS-Nummer: 180258-46-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR