Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Fmoc-trans-4-Amc-OH, trans-4-(((((9H-fluoren-9-yl)methoxy)carbonyl)amino)methyl)cyclohexanecarboxylic acid, N-Fmoc-tranexamic acid, trans-4-[[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]methyl]cyclohexanecarboxylic acid, trans-4-(Fmoc-aminomethyl)cyclohexanecarboxylic acid
OR52526-25G
APOSOR52526-25GEA
109
EUR
APOSOR52526-25G
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid
Formel:
C₂₃H₂₅NO₄ Molecular Weight: 379.46 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00273480 CAS-Nummer: 167690-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...