Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR52526-25G OR52526-5G
APOSOR52526-25GEA 0 EUR
APOSOR52526-25G APOSOR52526-5G
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid
Formel: C₂₃H₂₅NO₄
Lagertemperatur: Kühlschrank
MDL: MFCD00273480
CAS-Nummer: 167690-53-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR