(1S,2S)-1-Amino-2-indanol

Lieferant: Apollo Scientific
Achtung

Synonyme: (1S,2S)-1-Amino-2-indanol, (1S,2S)-(+)-1-Amino-2-hydroxyindan

OR52536-5G
APOSOR52536-5GEA 159 EUR
APOSOR52536-5G
(1S,2S)-1-Amino-2-indanol
(1S,2S)-(+)-1-Amino-2-indanol
Formel: C₉H₁₁NO
Molecular Weight: 149.19 g/mol
Schmelzpunkt: 142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08275383
CAS-Nummer: 163061-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR