Methyl 3-iodopyridine-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-iodopicolinate
OR52544-10G
APOSOR52544-10GEA
713
EUR
APOSOR52544-10G
Methyl 3-iodopyridine-2-carboxylate
Methyl 3-iodopyridine-2-carboxylate
Formel:
C₇H₆INO₂ Molecular Weight: 263.033 g/mol Schmelzpunkt: 39 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01860192 CAS-Nummer: 73841-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...