[3-(Methylsulphanyl)phenyl]hydrazine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR52548-1G
APOSOR52548-1GEA
604
EUR
APOSOR52548-1G
[3-(Methylsulphanyl)phenyl]hydrazine hydrochloride
[3-(Methylsulphanyl)phenyl]hydrazine hydrochloride
Formel:
C₇H₁₀N₂S·ClH Molecular Weight: 190,694 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12197080 CAS-Nummer: 940876-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...