[2-(Methylsulphanyl)phenyl]hydrazine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR52550-1G
APOSOR52550-1GEA 881 EUR
APOSOR52550-1G
[2-(Methylsulphanyl)phenyl]hydrazine hydrochloride
[2-(Methylsulphanyl)phenyl]hydrazine hydrochloride
Formel: C₇H₁₀N₂S·ClH
Molecular Weight: 190,694 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16990682
CAS-Nummer: 60481-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR