4-Hydrazinyl-6-methyl-2-(methylsulphanyl)pyrimidine
Lieferant: Apollo Scientific
OR52558-5G
APOSOR52558-5GEA
655
EUR
APOSOR52558-5G
4-Hydrazinyl-6-methyl-2-(methylsulphanyl)pyrimidine
4-Hydrazinyl-6-methyl-2-(methylsulphanyl)pyrimidine
Formel:
C₆H₁₀N₄S Molecular Weight: 170,235 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08691666 CAS-Nummer: 1980-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...