3-Cyclohexyl-L-alanine hydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-Amino-3-cyclohexylpropanoic acid hydrate, (S)-(+)-a-Aminocyclohexanepropionic acid hydrate

OR52577-5G
APOSOR52577-5GEA 52.2 EUR
APOSOR52577-5G
3-Cyclohexyl-L-alanine hydrate
3-Cyclohexyl-L-alanine hydrate
Formel: C₉H₁₇NO₂·H₂O
Molecular Weight: 189,252 g/mol
Schmelzpunkt: 234 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03703498
CAS-Nummer: 307310-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR