Boc-D-Ser(Me)-OH 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-2-((tert-Butoxycarbonyl)amino)-3-methoxypropanoic acid, Boc-O-Methyl-D-serine

OR52578-25G
APOSOR52578-25GEA 81.2 EUR
APOSOR52578-25G
Boc-D-Ser(Me)-OH 97%
Boc-D-Ser(Me)-OH
Formel: C₉H₁₇NO₅
Molecular Weight: 219.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08063987
CAS-Nummer: 86123-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR