1-Methylcyclopentan-1-amine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methylcyclopentanamine hydrochloride

OR52580-1G
APOSOR52580-1GEA 531 EUR
APOSOR52580-1G
1-Methylcyclopentan-1-amine hydrochloride
1-Methylcyclopentan-1-amine hydrochloride
Formel: C₆H₁₃N·ClH
Molecular Weight: 135,635 g/mol
Schmelzpunkt: 135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11858044
CAS-Nummer: 102014-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR