2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl
Lieferant: Apollo Scientific
|
Achtung
|
OR52584-10G
OR52584-5G
OR52584-25G
OR52584-100G
APOSOR52584-10GEA
0
EUR
APOSOR52584-10G
APOSOR52584-5G
APOSOR52584-25G
APOSOR52584-100G
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl
2-Di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl
Formel:
C₂₉H₄₅P Schmelzpunkt: 148…151 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06411306 CAS-Nummer: 564483-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...