Bis(pentamethylcyclopentadienyl)eisen(II)

Lieferant: Apollo Scientific
Achtung

Synonyme: Bis(pentamethylcyclopentadienyl)iron, Decamethylferrocene

OR52591-1G
APOSOR52591-1GEA 193 EUR
APOSOR52591-1G
Bis(pentamethylcyclopentadienyl)eisen(II)
Bis(pentamethylcyclopentadienyl)eisen(II)
Formel: C₂₀H₃₀Fe
Molecular Weight: 326,30 g/mol
Schmelzpunkt: 277 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00058736
CAS-Nummer: 12126-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR