Fmoc-Aad(otBu)-OH 95+%

Lieferant: Apollo Scientific
Achtung

OR52598-250MG
APOSOR52598-250MGEA 24.8 EUR
APOSOR52598-250MG
Fmoc-Aad(otBu)-OH 95+%
Fmoc-Aad(otBu)-OH
Formel: C₂₅H₂₉NO₆
Lagertemperatur: Raumtemperatur
MDL: MFCD00237006
CAS-Nummer: 159751-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR