5-Bromo-2,3-pyridinedicarboxylic anhydride

Lieferant: Apollo Scientific

OR52613-1G
APOSOR52613-1GEA 372 EUR
APOSOR52613-1G
5-Bromo-2,3-pyridinedicarboxylic anhydride
5-Bromo-2,3-pyridinedicarboxylic anhydride
Formel: C₇H₂BrNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD16250351
CAS-Nummer: 98278-78-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR