N,2-Dimethoxy-N-methylacetamide 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR52620-1G
APOSOR52620-1GEA
180
EUR
APOSOR52620-1G
N,2-Dimethoxy-N-methylacetamide 95%
N,2-Dimethoxy-N-methylacetamide
Formel:
C₅H₁₁NO₃ Molecular Weight: 133,146 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11848891 CAS-Nummer: 132289-57-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...