5-Methyluridin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

OR52639-100G
APOSOR52639-100GEA 98.6 EUR
APOSOR52639-100G
5-Methyluridin 97%
5-Methyluridin
Formel: C₁₀H₁₄N₂O₆
Molecular Weight: 258.23 g/mol
Schmelzpunkt: 183…184 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006535
CAS-Nummer: 1463-10-1
EINECS: 215-973-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR