Monosodium bromoisocyanurate hydrate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Bromoisocyanuric Acid Monosodium Salt
OR52663-25G
APOSOR52663-25GEA
177
EUR
APOSOR52663-25G
Monosodium bromoisocyanurate hydrate
Monosodium bromoisocyanurate hydrate
Formel:
C₃H₂BrN₃O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037229 CAS-Nummer: 164918-61-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...