H-Deg-OH

Lieferant: Apollo Scientific
Achtung

OR52678-1G OR52678-5G
APOSOR52678-1GEA 0 EUR
APOSOR52678-1G APOSOR52678-5G
H-Deg-OH
H-Deg-OH
Formel: C₆H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01737173
CAS-Nummer: 2566-29-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR