1-Methyl-1H-pyrazole-4,5-diamine sulphate 97%

Lieferant: Apollo Scientific
Achtung

OR52687-1G OR52687-250MG
APOSOR52687-1GEA 157 EUR
APOSOR52687-1G APOSOR52687-250MG
1-Methyl-1H-pyrazole-4,5-diamine sulphate 97%
1-Methyl-1H-pyrazole-4,5-diamine sulphate
Formel: C₄H₈N₄·H₂SO₄
Lagertemperatur: Kühlschrank
MDL: MFCD28099330
CAS-Nummer: 20055-01-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR