(-)-Benzotetramisole 98+%

Lieferant: Apollo Scientific

Synonyme: (-)-BTM, (S)-2-Phenyl-2,3-dihydrobenzo[d]imidazo[2,1-b]thiazole, (2S)-2,3-Dihydro-2-phenylimidazo[2,1-b]benzothiazole

OR52688-1G
APOSOR52688-1GEA 89.9 EUR
APOSOR52688-1G
(-)-Benzotetramisole 98+%
(-)-Benzotetramisole

Organocatalyst for the kinetic resolution of secondary alcohols by acylation with anhydrides.

Formel: C₁₅H₁₂N₂S
Schmelzpunkt: 95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD15072149
CAS-Nummer: 950194-37-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR