(1R,3S)-3-({[(2-Methyl-2-propanyl)oxy]carbonyl}amino)cyclohexancarbonsäure 97+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(1R,3S)-N-BOC-3-aminocyclohexanecarboxylic acid, (1R,3S)-3-((tert-Butoxycarbonyl)amino)cyclohexanecarboxylic acid, (1R,3S)-3-(Boc-amino)cyclohexanecarboxylic Acid
OR52690-1G
APOSOR52690-1GEA
707
EUR
APOSOR52690-1G
(1R,3S)-3-({[(2-Methyl-2-propanyl)oxy]carbonyl}amino)cyclohexancarbonsäure 97+%
(1R,3S)-3-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
Formel:
C₁₂H₂₁NO₄ Molecular Weight: 243,30 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973107 CAS-Nummer: 222530-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...