5-Bromo-2-hydroxy-4-methoxybenzaldehyde

Lieferant: Apollo Scientific

OR52694-1G
APOSOR52694-1GEA 77 EUR
APOSOR52694-1G
5-Bromo-2-hydroxy-4-methoxybenzaldehyde
5-Bromo-2-hydroxy-4-methoxybenzaldehyde
Formel: C₈H₇BrO₃
Molecular Weight: 231,043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00839421
CAS-Nummer: 57543-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR