3-Bromo-2-nitrobenzyl bromide 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-Bromo-3-(bromomethyl)-2-nitrobenzene

OR52712-1G OR52712-250MG OR52712-5G
APOSOR52712-1GEA 34.8 EUR
APOSOR52712-1G APOSOR52712-250MG APOSOR52712-5G
3-Bromo-2-nitrobenzyl bromide 95%
3-Bromo-2-nitrobenzyl bromide
Formel: C₇H₅Br₂NO₂
Molecular Weight: 294.928 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18390264
CAS-Nummer: 63471-25-0
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR