3-Bromo-2-nitrobenzyl bromide 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-Bromo-3-(bromomethyl)-2-nitrobenzene
OR52712-1G
OR52712-250MG
OR52712-5G
APOSOR52712-1GEA
34.8
EUR
APOSOR52712-1G
APOSOR52712-250MG
APOSOR52712-5G
3-Bromo-2-nitrobenzyl bromide 95%
3-Bromo-2-nitrobenzyl bromide
Formel:
C₇H₅Br₂NO₂ Molecular Weight: 294.928 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18390264 CAS-Nummer: 63471-25-0 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...