3-Bromo-2-iodobenzyl alcohol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-bromo-2-iodophenyl)methanol
OR52730-1G
OR52730-250MG
OR52730-5G
APOSOR52730-1GEA
201
EUR
APOSOR52730-1G
APOSOR52730-250MG
APOSOR52730-5G
3-Bromo-2-iodobenzyl alcohol 97%
3-Bromo-2-iodobenzyl alcohol
Formel:
C₇H₆BrIO Molecular Weight: 312.93 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18391655 CAS-Nummer: 1261571-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...