3-Bromo-2-iodobenzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-bromo-2-iodophenyl)methanol

OR52730-1G OR52730-250MG OR52730-5G
APOSOR52730-1GEA 201 EUR
APOSOR52730-1G APOSOR52730-250MG APOSOR52730-5G
3-Bromo-2-iodobenzyl alcohol 97%
3-Bromo-2-iodobenzyl alcohol
Formel: C₇H₆BrIO
Molecular Weight: 312.93 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18391655
CAS-Nummer: 1261571-03-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR