Methyl 5-oxohexanoate 98%

Lieferant: Apollo Scientific

OR52748-25G
APOSOR52748-25GEA 84.1 EUR
APOSOR52748-25G
Methyl 5-oxohexanoate 98%
Methyl 5-oxohexanoate
Formel: C₇H₁₂O₃
Siedepunkt: 100…101 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00038507
CAS-Nummer: 13984-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR