4-Bromo-2-piperidin-1-ylpyridine 96%

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-2-piperidinopyridine

OR52756-1G
APOSOR52756-1GEA 400 EUR
APOSOR52756-1G
4-Bromo-2-piperidin-1-ylpyridine 96%
4-Bromo-2-piperidin-1-ylpyridine
Formel: C₁₀H₁₃BrN₂
Molecular Weight: 241,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01646272
CAS-Nummer: 24255-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR