2-Bromo-3-(1-piperidyl)pyridine 95%
Lieferant: Apollo Scientific
OR52757-100MG
APOSOR52757-100MGEA
690
EUR
APOSOR52757-100MG
2-Bromo-3-(1-piperidyl)pyridine 95%
2-Bromo-3-(1-piperidyl)pyridine
Formel:
C₁₀H₁₃BrN₂ Molecular Weight: 241,128 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26405402 CAS-Nummer: 1563532-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...