1-(5-Bromo-3-pyridyl)piperazine 96%

Lieferant: Apollo Scientific

Synonyme: 1-(5-Bromopyridin-3-yl)piperazine

OR52760-1G
APOSOR52760-1GEA 177 EUR
APOSOR52760-1G
1-(5-Bromo-3-pyridyl)piperazine 96%
1-(5-Bromo-3-pyridyl)piperazine
Formel: C₉H₁₂BrN₃
Molecular Weight: 242,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09910230
CAS-Nummer: 412347-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR