3-Brom-5-(4-morpholino)pyridin 97%

Lieferant: Apollo Scientific

Synonyme: 4-(5-Bromo-3-pyridyl)morpholine, 3-Bromo-5-morpholinopyridine

OR52763-5G
APOSOR52763-5GEA 191 EUR
APOSOR52763-5G
3-Brom-5-(4-morpholino)pyridin 97%
3-Brom-5-(4-morpholino)pyridin
Formel: C₉H₁₁BrN₂O
Lagertemperatur: Kühlschrank
MDL: MFCD11656285
CAS-Nummer: 200064-13-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR