3-(1-Piperidyl)pyridin-4-amine 95%

Lieferant: Apollo Scientific

OR52770-100MG
APOSOR52770-100MGEA 2730 EUR
APOSOR52770-100MG
3-(1-Piperidyl)pyridin-4-amine 95%
3-(1-Piperidyl)pyridin-4-amine
Formel: C₁₀H₁₅N₃
Molecular Weight: 177,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20700896
CAS-Nummer: 144864-26-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR