5-Piperazin-1-ylpyridin-2-amine 99%

Lieferant: Apollo Scientific

Synonyme: 5-(Piperazin-1-yl)pyridin-2-amine

OR52774-250MG
APOSOR52774-250MGEA 242 EUR
APOSOR52774-250MG
5-Piperazin-1-ylpyridin-2-amine 99%
5-Piperazin-1-ylpyridin-2-amine
Formel: C₉H₁₄N₄
Molecular Weight: 178,234 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11604842
CAS-Nummer: 1082876-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR