3-Morpholinopyridin-2-amine 95%

Lieferant: Apollo Scientific

OR52777-250MG
APOSOR52777-250MGEA 609 EUR
APOSOR52777-250MG
3-Morpholinopyridin-2-amine 95%
3-Morpholinopyridin-2-amine
Formel: C₉H₁₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD11848423
CAS-Nummer: 1036645-86-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR