Methyl-3-brom-4-methoxybenzoat

Lieferant: Apollo Scientific

Synonyme: 3-Brom-4-methoxybenzoesäuremethylester

OR52901-25G
APOSOR52901-25GEA 195 EUR
APOSOR52901-25G
Methyl-3-brom-4-methoxybenzoat
Methyl-3-brom-4-methoxybenzoat
Formel: C₉H₉BrO₃
Molecular Weight: 245.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00210463
CAS-Nummer: 35450-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR