Methyl 4-chloro-2-methyl-pyrazole-3-carboxylate ≥95%

Lieferant: Apollo Scientific

OR52911-250MG OR52911-1G OR52911-5G
APOSOR52911-250MGEA 154 EUR
APOSOR52911-250MG APOSOR52911-1G APOSOR52911-5G
Methyl 4-chloro-2-methyl-pyrazole-3-carboxylate ≥95%
Methyl 4-chloro-2-methyl-pyrazole-3-carboxylate
Formel: C₆H₇ClN₂O₂
Molecular Weight: 174.58498 g/mol
MDL: MFCD02253778
CAS-Nummer: 400877-54-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR