Methyl-5-brom-2-chlorbenzoat 98%
Lieferant: Apollo Scientific
OR52912-100G
OR52912-25G
APOSOR52912-100GEA
513
EUR
APOSOR52912-100G
APOSOR52912-25G
Methyl-5-brom-2-chlorbenzoat 98%
Methyl-5-brom-2-chlorbenzoat
Formel:
C₈H₆BrClO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144770 CAS-Nummer: 251085-87-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...