Dimethyl (1,3-dihydro-3-oxo-1-isobenzofuranyl)phosphonate 98%

Lieferant: Apollo Scientific

Synonyme: Dimethyl (3-oxo-1,3-dihydroisobenzofuran-1-yl)phosphonate

OR52941-100G
APOSOR52941-100GEA 162 EUR
APOSOR52941-100G
Dimethyl (1,3-dihydro-3-oxo-1-isobenzofuranyl)phosphonate 98%
Dimethyl (1,3-dihydro-3-oxo-1-isobenzofuranyl)phosphonate
Formel: C₁₀H₁₁O₅P
Molecular Weight: 242.165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16883476
CAS-Nummer: 61260-15-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR