5-Bromo-6-chloro-1H-pyrazolo[3,4-b]pyridine 97%

Lieferant: Apollo Scientific

OR52942-250MG
APOSOR52942-250MGEA 464 EUR
APOSOR52942-250MG
5-Bromo-6-chloro-1H-pyrazolo[3,4-b]pyridine 97%
5-Bromo-6-chloro-1H-pyrazolo[3,4-b]pyridine
Formel: C₆H₃BrClN₃
Molecular Weight: 232,465 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29060202
CAS-Nummer: 1784380-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR