5-(5-Chlorosulphonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one 98%
Lieferant: Apollo Scientific
Synonyme:
4-Ethoxy-3-(1-methyl-7-oxo-3-propyl-6,7-dihydro-1H-pyrazolo[4,3-d]pyrimidin-5-yl)benzene-1-sulphonyl chloride
OR52944-5G
APOSOR52944-5GEA
206
EUR
APOSOR52944-5G
5-(5-Chlorosulphonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one 98%
5-(5-Chlorosulphonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
Formel:
C₁₇H₁₉ClN₄O₄S Molecular Weight: 410.875 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09753597 CAS-Nummer: 139756-22-2 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...