tert-Butyl N-(2-aminophenyl)-N-methylcarbamate

Lieferant: Apollo Scientific

Synonyme: tert-Butyl (2-aminophenyl)(methyl)carbamate, (2-Amino-phenyl)-methyl-carbamic acid tert-butyl ester

OR52955-1G
APOSOR52955-1GEA 778 EUR
APOSOR52955-1G
tert-Butyl N-(2-aminophenyl)-N-methylcarbamate
tert-Butyl N-(2-aminophenyl)-N-methylcarbamate
Formel: C₁₂H₁₈N₂O₂
Molecular Weight: 222,284 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05663971
CAS-Nummer: 885270-83-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR