3-Amino-3-oxopropanoic acid
Lieferant: Apollo Scientific
Synonyme:
MALONAMIC acid
OR52961-250MG
APOSOR52961-250MGEA
109
EUR
APOSOR52961-250MG
3-Amino-3-oxopropanoic acid
3-Amino-3-oxopropanoic acid
Formel:
C₃H₅NO₃ Molecular Weight: 103,08 g/mol Schmelzpunkt: 121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00154605 CAS-Nummer: 2345-56-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...