5-Nitro-1H-pyrazolo[3,4-b]pyridin-3-amine

Lieferant: Apollo Scientific
Achtung

OR52983-1G
APOSOR52983-1GEA 319 EUR
APOSOR52983-1G
5-Nitro-1H-pyrazolo[3,4-b]pyridin-3-amine
5-Nitro-1H-pyrazolo[3,4-b]pyridin-3-amine
Formel: C₆H₅N₅O₂
Molecular Weight: 179,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19690165
CAS-Nummer: 1186608-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR