1,8-Octylendiisothiocyanat 97%

Lieferant: Apollo Scientific

OR52987-1G
APOSOR52987-1GEA 109 EUR
APOSOR52987-1G
1,8-Octylendiisothiocyanat 97%
1,8-Octylendiisothiocyanat
Formel: C₁₀H₁₆N₂S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00041140
CAS-Nummer: 56312-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR