3-Oxa-2-thia-1-azaspiro[4.4]nonane 2,2-dioxide, N-BOC protected 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR52997-1G
OR52997-250MG
OR52997-5G
APOSOR52997-1GEA
145
EUR
APOSOR52997-1G
APOSOR52997-250MG
APOSOR52997-5G
3-Oxa-2-thia-1-azaspiro[4.4]nonane 2,2-dioxide, N-BOC protected 95%
3-Oxa-2-thia-1-azaspiro[4.4]nonane 2,2-dioxide, N-BOC protected
Formel:
C₁₁H₁₉NO₅S Lagertemperatur: Kühlschrank |
MDL:
MFCD31743878 CAS-Nummer: 2306264-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...